Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z256730

Kontes® threaded MICROFLEX® kits

Research kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Kimble®

Looking for similar products? Visit Product Comparison Guide

General description

Kit Components, Qty
A
Claisen adapter, 13-425 1
B Connecting adapter, 13-425 2
C Vacuum adapter, 13-425, 20-400 1
D Connector, 13-425 to Iin.-28 1
E Connector, 13-425 to hose connector 2
F Micro beaker, 10 mL 2
G Connector, 13-425 to 13-425 4
H Connector, 13-425 to 20-400 1
I Connector, 20-400 to 20-400 1
J Water-jacketed condenser, 13-425 1
K Drying tube/chromatography column, 2
50 mm, 13-425
L Air condenser/chromatography 1
column, 100 mm, 13-425
M Hickman distillation head 1
N PTFE filter, 30-60 μm, 13 mm 6
O PTFE filter support, 13 mm 6
P Cylindrical flask, 10 mL, 20-400 2
Q Micro vial, 1 mL, 13-425 2
R Micro vial, 3 mL, 20-400 2
S Micro vial, 5 mL, 20-400 2
T PTFE spin vane, 13 2
U PTFE spin vane, 20 2
V Micro spinbar, 2 × 7 mm 2
W Compression cap, 13-425 1
× Cap O-ring, Viton®, size 107 2
Foam insert set 1
Ekono-Case 1

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
MICROFLEX is a registered trademark of Kimble Kontes Asset Mgmt. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service