Key Documents
Z234230
Bend-A-Safety shield
H × W 24 in. × 48 in.
About This Item
UNSPSC Code:
46181702
NACRES:
NB.35
feature
splash goggles included
H × W
24 in. × 48 in.
General description
Flexible benchtop safety workshield. Constructed of 19 gauge 2x2 steel mesh core which fits into a translucent PVC film cover with a zip fastener along one edge. Freestanding; this shield can be formed into a variety of shapes. The mesh holds the shape while the vinyl cover provides protection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)