Key Documents
Z225622
Stainless steel forceps
Specimen, L 12 in.
About This Item
UNSPSC Code:
41122405
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 1 ea
L
12 in.
General description
with serrated tips
Application
Stainless steel forceps have been used for specimen embedding.
Features and Benefits
  • Available in different sizes, styles, and designs
  • Tips: pointed, blunt, rounded, curved, angled, or flat
  • Inner surfaces: plain or serrated for extra grip
  • Defined dimensions for precision work
  • Foam or silicone-covered tips for extra grip
  • Self-closing/locking guide pins to keep the tips aligned for a firmer grip
  • Inert polytetrafluoroethylene (PTFE) coating prevents contamination, scratching, or damage to delicate samples
Choose from one of the most recent versions:
Certificates of Analysis (COA)