Key Documents
Z224871
Sigma-Aldrich
Pinch clamps for SJ joints
Replacement pinch clamp for Aldrich combustion apparatus
About This Item
UNSPSC Code:
41122413
NACRES:
NB.43
material
stainless steel
Joint
joint SJ35/25
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
General description
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)