Key Documents
Z220590
Sigma-Aldrich
Solid-transfer vessel with two side-arms and a gas inlet with glass stopcock
capacity 75 mL, side arm male Joints: ST/NS 14/20 (2)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
label
U
Joint
male Joints: ST/NS side arm14/20 (2)
male joint ST/NS top45/40
capacity
75 mL
Related Categories
General description
For transferring and packaging air-sensitive solid compounds under inert atmosphere.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.