Key Documents
Z167517
Sigma-Aldrich
Aldrich® Schlenk-type Vigreux distilling column
male joint: ST/NS 14/20, female joint: ST/NS 14/20, L 150 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
label
O
Joint
female joint: ST/NS14/20
male joint: ST/NS14/20
L
150 mm
Related Categories
General description
Replacement part for Aldrich Air-Sensitive Glass kit. Thermoplastic screw caps with Viton O-rings usable to 140 deg C. Vigreux indents run the entire length of the column and act as the packing for the column.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.