Documents
Z188808
Supelco
Black plastic caps
Solid top, white rubber liner, thread for 8-425, pkg of 1000 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
black phenolic cap
white rubber liner
feature
closure type black phenolic solid top cap
packaging
pkg of 1000 ea
NMR tube diam.
5 mm
fitting
thread for 8-425
General description
It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.
These autoclavable caps are designed to ensure the ultimate in product safety.