Key Documents
Z130974
Nalgene® Griffin beakers
capacity 250 mL, translucent polypropylene
Synonym(s):
PP beaker, nalgene PP beaker
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
translucent polypropylene
feature
graduations
packaging
pkg of 6 ea
manufacturer/tradename
Nalgene 1201-0250
capacity
250 mL
General description
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Other Notes
Browse all plastic beakers
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)