Key Documents
Y906603
Sigma-Aldrich
Valve
brass diaphragm, for CGA 110/170 right angle path, internal/external, right hand thread
About This Item
UNSPSC Code:
12142200
NACRES:
NA.12
material
brass diaphragm
parameter
3000 psi max. pressure
L
3.31 in.
weight
230 g
fitting
for CGA 110/170 right angle path, internal/external, right hand thread
Other Notes
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Choose from one of the most recent versions:
Certificates of Analysis (COA)