Key Documents
WHAWB100029
Unicore Punch Kit
I.D. 2.0 mm, pkg of 4 ea
Synonym(s):
UniCore Punch Kit 2.00 mm (4), Whatman FTA, Z747378, cutting mat, harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
Pricing and availability is not currently available.
packaging
pkg of 4 ea
manufacturer/tradename
Qiagen WB100029
I.D.
2.0 mm
General description
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Manual UniCore punch system for sample disc removal from FTA cards.
A cutting mat (also available separately: WHAWB100022) included in UniCore Punch Kits ensures clean sample cuts from FTA/FTA Elute Cards and extends the life time of the cutting tip (up to 500 punches).
Features and Benefits
  • Stainless steel cutting edges Stainless steel cutting edges (2.0 mm) enable manual coring, retrieval, and controlled ejection of the FTA punch.
  • Designed to minimize contamination and carryover between samples when a cleaning procedure is followed.
Other Notes
Field of Use : For internal research use only
Legal Information
FTA is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Articles
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Protocols
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.