Key Documents
WHAWB120462
EasiCollect (50)
device for collection of buccal DNA samples, pkg of pack of 50 devices
Synonym(s):
Whatman FTA
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
feature
Included: A removable 2 × 2 in. (5 × 5 cm) indicating FTA card.
manufacturer/tradename
Qiagen WB120462
packaging
pkg of pack of 50 devices
General description
EasiCollect device simplifies buccal cell collection and transportation. The design has been engineered for maximal cell collection and uniform sample application to FTA Cards to overcome individual differences. The pressure clips molded into the EasiCollect device ensure a uniform and constant pressure for reproducible transfer of cells to the FTA Card.
Features and Benefits
50 buccal sample collection devices for simplified sample uptake and transportation.
Other Notes
Field of Use : For internal research use only
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.