Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA67063602

Whatman® Polycap AS encapsulated filter for aqueous solutions

Polycap AS 36 with filling bell, pore size 0.2 μm, stepped barb inlet, stepped barb outlet

Synonym(s):

Whatman filter, Z743844, in line filters, syringe filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

material

Nylon membrane
polypropylene housing
stepped barb inlet
stepped barb outlet

sterility

sterile

packaging

pkg of 1 ea

manufacturer/tradename

Whatman 6706-3602
Whatman Article No. 28420142 (US reference)

diam.

36 mm

pore size

0.2 μm

Looking for similar products? Visit Product Comparison Guide

General description

Whatman Polycap AS (aqueous solutions) capsule filters from Cytiva combines a glass microfiber (GMF) prefilter over a nylon membrane final filter to provide maximum filtration of difficult, particulate-laden solutions.

Polycap AS for aqueous solutions

Polycap AS are Whatman capsule filters specifically designed for high-volume filtration of aqueous sample solutions in a compact size. With a nylon membrane filter and GMF prefilter, these filters have a long usable life.

Application

Biologicals, Buffers, Cleaning/rinsing solutions, Enzymes, Extemporaneous solutions, Immunologicals, Irrigation solutions, Nutrients, Pharmaceutical solutions, Reagent preparation, Salt solutions, Therapy solutions, Tissue culture media, Viral suspensions.

Features and Benefits

  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 µm) life and higher filtration efficiency.
  • Nylon membrane filter layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates
  • Polypropylene housing is thermally fused (no glues, adhesives, or extraneous materials), reducing extractable levels
  • Can be tested with bubble point, pressure decay, or forward flow methods
  • Available in presterilized and nonsterile (autoclavable) versions
  • Manufactured according to ISO9001:2008 QMS standards
  • Recyclable after use.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service