Key Documents
WHA10442101
Whatman® glass vacuum filtration device
GV050 series, GV050/1/03 support screen
Synonym(s):
Z750859
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
borosilicate glass funnel(s) (funnel)
stainless steel clamp(s) (clamp)
packaging
pk of 1
manufacturer/tradename
Whatman 10442101
Whatman Article No. 28415562 (US reference)
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
General description
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Features and Benefits
  • Accessory to GV050 Glass Filter Funnel
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva