Key Documents
WAMUPG2HCPWC1
Whatman® Mini-UniPrep® G2 hand compressor
1 position, pack of 1
Synonym(s):
hand compressor, mini-uniprep G2, syringeless filter, whatman filter
About This Item
UNSPSC Code:
40161807
NACRES:
NB.24
packaging
pack of 1
manufacturer/tradename
Whatman Article No. 29026195 (US reference)
Whatman MUPG2HCPWC1
General description
Mini-UniPrep G2 Hand Compressor is for use with Mini-UniPrep G2 Syringeless filters.
Features and Benefits
  • Ergonomic hand compressor allows for sample preparation with increased hand comfort
  • Mini-UniPrepAll-in-One syringeless filter prepares samples three times faster than standard syringe filter method
  • Mini-UniPrepglass or polypropylene vials support analyses such as pharmaceutical quality control (pharma QC) and water monitoring
  • Mini-UniPrepvials can be loaded directly into the autosampler
Legal Information
Mini-UniPrep is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)