Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W6799

Pyruvic-1,2-13C2 acid

API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)

Synonym(s):

2-Oxo-propanoic-1,2-13C2 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CO13CO2H
CAS Number:
Molecular Weight:
90.05
MDL number:
UNSPSC Code:
12352115

grade

cGMP

isotopic purity

≥99 atom % 13C

Assay

≥95% (CP)

bp

165 °C (lit.)

mass shift

M+2

storage temp.

-10 to -25°C

SMILES string

C[13C](=O)[13C](O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)/i2+1,3+1

InChI key

LCTONWCANYUPML-SUEIGJEOSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service