Skip to Content
MilliporeSigma
Pyruvic-1,2-13C2 acid API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)

Pyruvic-1,2-13C2 acid

Pyruvic-1,2-13C2 acid API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)
Synonym(s):
2-Oxo-propanoic-1,2-13C2 acid
Linear Formula:
CH313CO13CO2H
CAS Number:
Molecular Weight:
90.05

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Special Grade