Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W403622

Sigma-Aldrich

cis-4-Dodecenal

≥94%, stabilized, FG

Synonym(s):

(Z)-4-Dodecenal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
FEMA Number:
4036
UNSPSC Code:
12164502
Flavis number:
05.220
NACRES:
NA.21
Organoleptic:
citrus; waxy; fruity
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥94%

contains

synthetic α-tocopherol as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

citrus; waxy; fruity

SMILES string

O=CCCC=CCCCCCCC

InChI

1S/C12H22O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h8-9,12H,2-7,10-11H2,1H3

InChI key

MMCDSVCBSAMNPL-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service