Skip to Content
MilliporeSigma
All Photos(1)

Documents

W358002

Sigma-Aldrich

cis-6-Nonenal

≥95%, FG

Synonym(s):

(Z)-non-6-enal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CH(CH2)4CHO
CAS Number:
Molecular Weight:
140.22
FEMA Number:
3580
EC Number:
Council of Europe no.:
661
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.059
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥95%

refractive index

n20/D 1.442 (lit.)

bp

87 °C/19 mmHg (lit.)

density

0.841 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cucumber; green; melon; waxy

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCC\C([H])=C(\[H])CC

InChI

1S/C9H16O/c1-2-3-4-5-6-7-8-9-10/h3-4,9H,2,5-8H2,1H3/b4-3-

InChI key

RTNPCOBSXBGDMO-ARJAWSKDSA-N

Application


  • Anti-Methanogenic Traits of Safflower Oil Compounds Against Methyl-Coenzyme M Reductase Receptor in Equines: An In Silico Docking Analysis.: This study explores the potential of cis-6-Nonenal as a compound in safflower oil that exhibits anti-methanogenic properties. The in silico docking analysis highlights its application in reducing methane emissions in equines, contributing to environmental sustainability and agricultural practices (Khusro et al., 2022).

  • The formation of highly oxidized multifunctional products in the ozonolysis of cyclohexene.: This research investigates the role of cis-6-Nonenal in the formation of highly oxidized multifunctional products during the ozonolysis of cyclohexene. The study provides insights into atmospheric chemistry and the processes involved in the formation of secondary organic aerosols, which have implications for air quality and climate change (Rissanen et al., 2014).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

P W Smith et al.
Journal of dairy science, 67(4), 794-801 (1984-04-01)
Solvent extraction and nitrogen degassing were employed to collect volatile odors from the lipid complex yielded by swabbing the sebaceous gland areas of the scalps of sexually active bucks from four breeds of goats. Gland tissue samples from freshly slaughtered
Cristina Avonto et al.
Toxicology and applied pharmacology, 289(2), 177-184 (2015-10-13)
Skin sensitization is an important toxicological end-point in the risk assessment of chemical allergens. Because of the complexity of the biological mechanisms associated with skin sensitization, integrated approaches combining different chemical, biological and in silico methods are recommended to replace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service