Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W347507

Sigma-Aldrich

Trithioacetone

≥99%, FG

Synonym(s):

2,2,4,4,6,6-Hexamethyl-s-trithiane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18S3
CAS Number:
Molecular Weight:
222.43
FEMA Number:
3475
EC Number:
Council of Europe no.:
2334
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.009
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.54 (lit.)

bp

105-107 °C/10 mmHg (lit.)

mp

24 °C (lit.)

density

1.065 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; burnt; green; earthy; sulfurous

SMILES string

CC1(C)SC(C)(C)SC(C)(C)S1

InChI

1S/C9H18S3/c1-7(2)10-8(3,4)12-9(5,6)11-7/h1-6H3

InChI key

NBNWHQAWKFYFKI-UHFFFAOYSA-N

Biochem/physiol Actions

Taste at 0.35-0.50 ppm

Other Notes

Natural occurrence: Roast beef.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 24, 42-42 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service