Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W358126

Sigma-Aldrich

3-Octanol

greener alternative

natural, ≥97%, FCC, FG

Synonym(s):

Ethyl pentyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH2CH3
CAS Number:
Molecular Weight:
130.23
FEMA Number:
3581
Beilstein:
1719310
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.098
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

~4.5 (vs air)

vapor pressure

~1 mmHg ( 20 °C)

Assay

≥97%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.426 (lit.)

bp

174-176 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cheese; creamy; mushroom; musty; earthy

SMILES string

CCCCCC(O)CC

InChI

1S/C8H18O/c1-3-5-6-7-8(9)4-2/h8-9H,3-7H2,1-2H3

InChI key

NMRPBPVERJPACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Octanol is one of the major volatile flavor components of mushroom.
Natural occurrence: Spearmint oil, oat groats, Ocimum basilicum, pimento leaf and truffle.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application

  • Impact of aroma-enhancing microorganisms on aroma attributes of industrial Douchi: An integrated analysis using E-nose, GC-IMS, GC-MS, and descriptive sensory evaluation.: This study explores the enhancement of aroma attributes in Douchi through the use of aroma-enhancing microorganisms. It highlights the role of 3-Octanol in modifying flavor profiles, which could lead to improved food processing techniques (Li et al., 2024).

  • Nematicidal Activity of Phytochemicals against the Root-Lesion Nematode Pratylenchus penetrans.: Investigates the potential use of 3-Octanol as a natural nematicide. This research highlights its effectiveness in controlling agricultural pests, offering a safer alternative to synthetic pesticides (Barbosa et al., 2024).

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mushroom flavor.
Maga JA.
Journal of Agricultural and Food Chemistry, 29(1), 1-4 (1981)
Volatile composition of fourteen species of fresh wild mushrooms (Boletales).
Rapior S, et al.
J. Essent. Oil Res., 9(2), 231-234 (1997)
Nannan Chen et al.
Biochemical and biophysical research communications, 460(4), 944-948 (2015-04-07)
Age-related memory impairment (AMI) is a phenomenon observed from invertebrates to human. Memory extinction is proposed to be an active inhibitory modification of memory, however, whether extinction is affected in aging animals remains to be elucidated. Employing a modified paradigm
Yichun Shuai et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(48), E6663-E6672 (2015-12-03)
Recent studies have identified molecular pathways driving forgetting and supported the notion that forgetting is a biologically active process. The circuit mechanisms of forgetting, however, remain largely unknown. Here we report two sets of Drosophila neurons that account for the
Mosciano, G.
Perfumer & Flavorist, 21, 53-53 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service