Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W358118

Sigma-Aldrich

3-Octanol

≥97%, FCC, FG

Synonym(s):

Ethyl pentyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH2CH3
CAS Number:
Molecular Weight:
130.23
FEMA Number:
3581
Beilstein:
1719310
EC Number:
Council of Europe no.:
4023
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.098
NACRES:
NA.21
Organoleptic:
creamy; mushroom; musty; earthy; minty; waxy
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

~4.5 (vs air)

vapor pressure

~1 mmHg ( 20 °C)

Assay

≥97%

refractive index

n20/D 1.426 (lit.)

bp

174-176 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; mushroom; musty; earthy; minty; waxy

SMILES string

CCCCCC(O)CC

InChI

1S/C8H18O/c1-3-5-6-7-8(9)4-2/h8-9H,3-7H2,1-2H3

InChI key

NMRPBPVERJPACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • RIFM fragrance ingredient safety assessment, 3-octanol, CAS Registry Number 589-98-0: This safety assessment review offers a thorough examination of 3-Octanol′s application in the fragrance industry, discussing its solvent properties, safety, and handling in laboratory settings, thus serving as a critical reference for researchers focused on industrial applications and safety of fragrance ingredients (Api et al., 2021).

Biochem/physiol Actions

Taste at 10 ppm

Other Notes

Natural occurrence: Spearmint oil, oat groats, Ocimum basilicum, pimento leaf and truffle.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nannan Chen et al.
Biochemical and biophysical research communications, 460(4), 944-948 (2015-04-07)
Age-related memory impairment (AMI) is a phenomenon observed from invertebrates to human. Memory extinction is proposed to be an active inhibitory modification of memory, however, whether extinction is affected in aging animals remains to be elucidated. Employing a modified paradigm
Mosciano, G.
Perfumer & Flavorist, 21, 53-53 (1996)
Yichun Shuai et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(48), E6663-E6672 (2015-12-03)
Recent studies have identified molecular pathways driving forgetting and supported the notion that forgetting is a biologically active process. The circuit mechanisms of forgetting, however, remain largely unknown. Here we report two sets of Drosophila neurons that account for the
Elena Canellas et al.
Journal of mass spectrometry : JMS, 49(11), 1181-1190 (2014-11-15)
Pressure-sensitive adhesives (PSA) are used to manufacture labels that are applied directly on the food. These adhesives could contain not only intentionally added compounds (IAS) to the adhesive formula but also non-intentionally added substances (NIAS), due to the impurities from
Mirjam Appel et al.
PloS one, 10(5), e0126986-e0126986 (2015-05-21)
Electric shock is a common stimulus for nociception-research and the most widely used reinforcement in aversive associative learning experiments. Yet, nothing is known about the mechanisms it recruits at the periphery. To help fill this gap, we undertook a genome-wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service