Skip to Content
MilliporeSigma
All Photos(1)

Documents

W324000

Sigma-Aldrich

o-Toluenethiol

≥97%, FG

Synonym(s):

2-Methylbenzenethiol, o-Thiocresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SH
CAS Number:
Molecular Weight:
124.20
FEMA Number:
3240
EC Number:
Council of Europe no.:
2272
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.027
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.578 (lit.)

bp

195 °C (lit.)

mp

10-12 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bacon; meaty; sulfurous

SMILES string

Cc1ccccc1S

InChI

1S/C7H8S/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

LXUNZSDDXMPKLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • RIFM fragrance ingredient safety assessment, o-toluenethiol, CAS Registry Number 137-06-4.: This study conducted a safety assessment of o-toluenethiol as a fragrance ingredient.(Api et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F

Flash Point(C)

63 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service