Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W291706

Sigma-Aldrich

Polysorbate 80

FG

Synonym(s):

TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2917
MDL number:
UNSPSC Code:
12164502
E Number:
E 433
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
palm oil
synthetic
food allergen:
corn
Pricing and availability is not currently available.

biological source

palm oil
synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 172.515
FDA 21 CFR 172.836
FDA 21 CFR 172.838
FDA 21 CFR 172.840
FDA 21 CFR 172.842

vapor pressure

<1 mmHg ( 20 °C)

refractive index

n20/D 1.472 (lit.)

density

1.064 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

corn

Organoleptic

alcohol

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polysorbate 80 (TWEEN® 80) is a polyethylene sorbitol ester.

Application

Polysorbate 80 (TWEEN® 80), a hydrophilic non-ionic surfactant,[1] is widely employed as a surfactant and solubilizing agent in food and cosmetic industries.[2]

Legal Information

TWEEN is a registered trademark of Croda International PLC

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongjian Zhang et al.
Archives of pharmacal research, 26(9), 768-772 (2003-10-17)
Tween 80 (Polysorbate 80) is a hydrophilic nonionic surfactant commonly used as an ingredient in dosing vehicles for pre-clinical in vivo studies (e.g., pharmacokinetic studies, etc.). Tween 80 increased apical to basolateral permeability of digoxin in Caco-2 cells suggesting that
Stability characterization of a polysorbate 80-dimethyl trisulfide formulation, a cyanide antidote candidate.
Bartling CM, et al.
Drugs in R&D, 16(1), 109-127 (2016)
Use of Polysorbate 80 (Tween 80) as a phase-transfer catalyst.
Thoman CJ, et al.
The Journal of Organic Chemistry, 54(18), 4476-4478 (1989)
Tatiana Beldarrain-Iznaga et al.
Food research international (Ottawa, Ont.), 141, 110136-110136 (2021-03-02)
To provide their health effect, probiotics need to maintain their viability, adhere to the intestinal epithelium, and colonize it without losing their probiotic properties. In the present study, Lactobacillus casei was encapsulated in a double emulsion and then coated with
Asher Preska Steinberg et al.
Biomacromolecules, 20(7), 2675-2683 (2019-06-28)
Systems consisting of a polyelectrolyte solution in contact with a cross-linked polyelectrolyte network are ubiquitous (e.g., biofilms, drug-delivering hydrogels, and mammalian extracellular matrices), yet the underlying physics governing these interactions is not well understood. Here, we find that carboxymethyl cellulose

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Storage conditions of Polysorbate 80: What are the adviced storage conditions of Polysorbate 80? We normally store it at room temperature. But recently stored it at 2-8°C, does this impact the quality of our product?

    1 answer
    1. This product does not require controlled temperature storage. While stability studies at low temperatures have not been performed, historically, refrigerated temperatures have had no impact on product integrity. In many cases, this product is added to mixtures at low concentrations and then stored frozen. This product ships at ambient temperature globally, at all times of the year, and in all climates. It is very robust.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service