Skip to Content
MilliporeSigma

9490-OP

Polysorbate 80

OmniPur®

Synonym(s):

TWEEN® 80, Polyoxyethylene (20) Sorbitan Monooleate, Polyoxyethylene Sorbitan Monooleate, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

viscous liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

gold-yellow

shipped in

ambient

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

General description

Non-ionic detergent suited for solubilization of membrane proteins, enzymology, analytical, and diagnostic, work as well as chromatography and electrophoresis. Molecular Biology Grade.

DNAse: none detected
Heavy metals (as Pb): ≤0.001%
Hydroxyl number: 65-80
Residue after ignition: ≤0.25%
Moisture (H2O): ≤3%
Protease: none detected
RNAse: none detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Surfactants′ Interplay with Biofilm Development in Staphylococcus and Candida.: This study explores how surfactants, including Polysorbate 80, influence biofilm development in Staphylococcus and Candida species. The findings highlight the critical role of surfactants in modulating biofilm formation, which has implications for developing new therapeutic strategies against biofilm-associated infections. (Aonofriesei, 2024).

  • Stress-mediated polysorbate 20 degradation and its potential impact on therapeutic proteins.: This article examines the degradation of Polysorbate 20 under stress conditions and its effects on therapeutic proteins. While primarily focusing on Polysorbate 20, the insights are relevant for understanding the stability and formulation challenges of similar surfactants like Polysorbate 80 in pharmaceutical applications. (Aryal et al., 2024).

  • Dissecting the toxicological profile of polysorbate 80 (PS80): comparative analysis of constituent variability and biological impact using a zebrafish model.: This comprehensive study evaluates the toxicological effects of Polysorbate 80 using a zebrafish model, providing detailed insights into its safety profile and biological impact, which are crucial for its use in pharmaceuticals and other applications. (Wang et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Toxicity: Irritant (B)


Still not finding the right product?

Explore all of our products under Polysorbate 80


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

>235.4 °F - closed cup

flash_point_c

> 113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library