Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W291218

Sigma-Aldrich

Piperonyl acetate

≥99%, FG

Synonym(s):

heliotropyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
FEMA Number:
2912
EC Number:
Council of Europe no.:
2068
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.220
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.5265 (lit.)

bp

150-151 °C/10 mmHg (lit.)

density

1.227 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; sweet

SMILES string

CC(=O)OCc1ccc2OCOc2c1

InChI

1S/C10H10O4/c1-7(11)12-5-8-2-3-9-10(4-8)14-6-13-9/h2-4H,5-6H2,1H3

InChI key

PFWYHTORQZAGCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S358-S362 (2012-03-27)
A toxicologic and dermatologic review of piperonyl acetate when used as a fragrance ingredient is presented. Piperonyl acetate is a member of the fragrance structural group aryl alkyl alcohol simple acid esters (AAASAE). The AAASAE fragrance ingredients are prepared by
D J Wright et al.
Xenobiotica; the fate of foreign compounds in biological systems, 10(4), 265-270 (1980-04-01)
1. The metabolism of piperonylidene acetone, and the food additives piperonyl acetone, piperonyl acetate and piperonyl isobutyrate were studied in male rabbits after oral doses of 100 mg/kg. 2. Both piperonylidene acetone and piperonyl acetone were extensively excreted as 4-(3,4-methylenedioxyphenyl)-butan-2-ol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service