Skip to Content
MilliporeSigma
All Photos(1)

Documents

166200

Sigma-Aldrich

Furfuryl acetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

175-177 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCc1ccco1

InChI

1S/C7H8O3/c1-6(8)10-5-7-3-2-4-9-7/h2-4H,5H2,1H3

InChI key

CKOYRRWBOKMNRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Furfuryl acetate was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Mehner et al.
Molecules (Basel, Switzerland), 12(3), 634-640 (2007-09-14)
5-Acetoxymethyl- and 5-hydroxymethyl-2-vinylfuran were synthesized by two routes. The first route starts from 2-methylfuran and the second from furfuryl acetate. The latter route, involving successive Vilsmeier-Haack and Wittig reactions, is suitable for producing 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2 vinylfuran in 68% and
R Daniel Cacciari et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 16(11), 1717-1726 (2017-10-27)
The use of biologically active substances with anti-inflammatory properties such as corticosteroids has increased considerably in the last few decades. Particularly, the compound we are interested in, prednisolone (Predn), is a glucocorticoid with high biological activity. This compound absorbs UV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service