Skip to Content
MilliporeSigma
All Photos(1)

Documents

W287806

Sigma-Aldrich

Phenylacetic acid

≥99%, FCC, FG

Synonym(s):

α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2H
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2878
Beilstein:
1099647
EC Number:
Council of Europe no.:
672
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.038
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

~4 (vs air)

vapor pressure

1 mmHg ( 97 °C)

Assay

≥99%

bp

265 °C (lit.)

mp

76-78 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; honey; floral; sweet

SMILES string

OC(=O)Cc1ccccc1

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

WLJVXDMOQOGPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phenylacetic acid is a volatile acid that is reported to occur in balsamic vinegar and cocoa powder.

Application


  • The Phenylacetic Acid Catabolic Pathway Regulates Antibiotic and Oxidative Stress Responses in Acinetobacter.: This study details the phenylacetic acid catabolic pathway in Acinetobacter and its regulatory roles in antibiotic resistance and oxidative stress response, highlighting its biochemical significance (Hooppaw et al., 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin-layer high-vacuum distillation to isolate volatile flavour compounds of cocoa powder.
Krings U, et al.
European Food Research and Technology, 223(5), 675-675 (2006)
Solid?phase extraction method for determination of volatile compounds in traditional balsamic vinegar.
Duran Guerrero E, et al.
Journal of Separation Science, 31(16?17), 3030-3036 (2008)
Gustav Nyström et al.
Nature communications, 6, 7259-7259 (2015-05-30)
Traditional thin-film energy-storage devices consist of stacked layers of active films on two-dimensional substrates and do not exploit the third dimension. Fully three-dimensional thin-film devices would allow energy storage in bulk materials with arbitrary form factors and with mechanical properties
D C Singleton et al.
Oncogene, 34(36), 4713-4722 (2014-12-09)
Hypoxia is a common feature of locally advanced breast cancers that is associated with increased metastasis and poorer survival. Stabilisation of hypoxia-inducible factor-1α (HIF1α) in tumours causes transcriptional changes in numerous genes that function at distinct stages of the metastatic
L Ren et al.
Nanotechnology, 26(28), 284001-284001 (2015-06-27)
The prospect for spatial imaging with mass spectroscopy at the level of the cell requires new means of cell extraction to conserve molecular structure. To this aim, we demonstrate a new laser extraction process capable of extracting intact biological entities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service