Skip to Content
MilliporeSigma
Phenylacetic acid British Pharmacopoeia (BP) Reference Standard

Phenylacetic acid

Phenylacetic acid British Pharmacopoeia (BP) Reference Standard
Synonym(s):
α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA, Phenylacetic acid
Linear Formula:
C6H5CH2CO2H
CAS Number:
Molecular Weight:
136.15

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique