Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W277126

Sigma-Aldrich

Neroli oil, Morocco

FG

Synonym(s):

Citrus bigaradia oil morocco, Neroli marocco EO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2771
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
orange
grade:
FG
Halal
Kosher
biological source:
Citrus aurantium L.
food allergen:
no known allergens

biological source

Citrus aurantium L.

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 182.20

origin

France origin

refractive index

n20/D 1.47 (lit.)

bp

163 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange

Preparation Note

Extraction method: steam distillation

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service