Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W311936

Sigma-Aldrich

Ylang-ylang oil

extra, blend, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3119
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
Cananga odorata
synthetic
food allergen:
no known allergens

biological source

Cananga odorata
synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 182.20

optical activity

[α]/D -12.0 to -17.5°

composition

contains IFRA restricted ylang-ylang oil

refractive index

n20/D 1.503

density

0.963 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

geraniol, eugenol, farnesol, Cananga odorata and ylang-ylang oil, linalyl acetate, isoeugenol, benzyl alcohol, linalool, β-caryophyllene (ox.), benzyl salicylate, benzyl benzoate

Organoleptic

floral

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service