Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W274208

Sigma-Aldrich

Methyl propionate

≥98%, FG

Synonym(s):

Methyl propanoate, Propanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOCH3
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2742
Beilstein:
1737628
EC Number:
Council of Europe no.:
415
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.134
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

vapor density

3 (vs air)

vapor pressure

40 mmHg ( 11 °C)
85.3 mmHg ( 25 °C)

Assay

≥98%

autoignition temp.

876 °F

expl. lim.

13 %

refractive index

n20/D 1.376 (lit.)

bp

79 °C (lit.)

mp

−88 °C (lit.)

density

0.915 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; fruity; strawberry

SMILES string

CCC(=O)OC

InChI

1S/C4H8O2/c1-3-4(5)6-2/h3H2,1-2H3

InChI key

RJUFJBKOKNCXHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ileana M Pazos et al.
Angewandte Chemie (International ed. in English), 53(24), 6080-6084 (2014-05-03)
The ability to quantify the local electrostatic environment of proteins and protein/peptide assemblies is key to gaining a microscopic understanding of many biological interactions and processes. Herein, we show that the ester carbonyl stretching vibration of two non-natural amino acids
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service