MilliporeSigma
  • Ester carbonyl vibration as a sensitive probe of protein local electric field.
Ester carbonyl vibration as a sensitive probe of protein local electric field.
Angewandte Chemie (International ed. in English) (2014-05-03)
Ileana M Pazos, Ayanjeet Ghosh, Matthew J Tucker, Feng Gai
ABSTRACT
The ability to quantify the local electrostatic environment of proteins and protein/peptide assemblies is key to gaining a microscopic understanding of many biological interactions and processes. Herein, we show that the ester carbonyl stretching vibration of two non-natural amino acids, L-aspartic acid 4-methyl ester and L-glutamic acid 5-methyl ester, is a convenient and sensitive probe in this regard, since its frequency correlates linearly with the local electrostatic field for both hydrogen-bonding and non-hydrogen-bonding environments. We expect that the resultant frequency-electric-field map will find use in various applications. Furthermore, we show that, when situated in a non-hydrogen-bonding environment, this probe can also be used to measure the local dielectric constant (ε). For example, its application to amyloid fibrils formed by Aβ(16-22) revealed that the interior of such β-sheet assemblies has an ε value of approximately 5.6.
Sigma-Aldrich
Methyl propionate, 99%
Sigma-Aldrich
Methyl propionate, ≥98%, FG
Supelco
Methyl propionate, analytical standard
Sigma-Aldrich
Methyl acetate, ≥98%, FG
Sigma-Aldrich
Methyl acetate, anhydrous, 99.5%
Supelco
Methyl acetate, analytical standard
Sigma-Aldrich
Methyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Sigma-Aldrich
Methyl acetate, ReagentPlus®, 99%
Sigma-Aldrich
Methyl acetate, suitable for HPLC, ≥99.8%