W272108
Methyl 4-methylvalerate
≥97%
Synonym(s):
Methyl isocaproate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH2CH2CO2CH3
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2721
EC Number:
Council of Europe no.:
322
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.432
NACRES:
NA.21
Recommended Products
biological source
synthetic
Quality Level
Assay
≥97%
refractive index
n20/D 1.404 (lit.)
bp
139-140 °C (lit.)
density
0.888 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Organoleptic
banana; fruity; pineapple; sweet
SMILES string
COC(=O)CCC(C)C
InChI
1S/C7H14O2/c1-6(2)4-5-7(8)9-3/h6H,4-5H2,1-3H3
InChI key
KBCOVKHULBZKNY-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
102.2 °F - closed cup
Flash Point(C)
39 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service