Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

639079

Sigma-Aldrich

3-Methyl-2-buten-2-ylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2)2C=CCH3BO2
CAS Number:
Molecular Weight:
113.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

173-178 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(C)=C(\C)B(O)O

InChI

1S/C5H11BO2/c1-4(2)5(3)6(7)8/h7-8H,1-3H3

InChI key

ONHWZTRGTFWBIX-UHFFFAOYSA-N

Application

Reactant involved in:
  • Synthesis of 8-alkenylboronpyrromethene dyes
  • Addition reactions with N-methyliminodiacetic acid or aromatic, heteroaromatic or aliphatic aldehydes
  • Arylation of methyl o-formylbenzoates
  • Synthesis of tert-butanesulfinyl α-branched allylic amines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service