Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W208620

Sigma-Aldrich

Anethol

natural, 99%, FG

Synonym(s):

trans-Anethole, 4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
FEMA Number:
2086
Beilstein:
774229
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.010
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

99%

composition

contains trans-Anethole

refractive index

n20/D 1.561 (lit.)

bp

234-237 °C (lit.)

mp

20-21 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

trans-anethole

Organoleptic

anise; sweet

SMILES string

COc1ccc(\C=C\C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Naturally occurring phenylpropene derivative that is estrogenic at lower concentrations and cytotoxic at higher concentrations to cancer cell lines. The cytotoxicity is related to the metabolism of trans-anethole to 4-hydroxy-1-propenylbenzene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riham Gharib et al.
Food chemistry, 267, 67-74 (2018-06-24)
The effect of hydrogenation of phospholipids on the characteristics of freeze-dried liposomes was investigated using hydroxypropyl-ß-cyclodextrin (HP-ß-CD) as membrane protectant. The ethanol-injection method was applied to prepare liposomes using hydrogenated (Phospholopion-90H and 80H) and non-hydrogenated phospholipids (Lipoid-S100) in combination with
Riham Gharib et al.
The Journal of membrane biology, 250(3), 259-271 (2017-04-24)
The effect of structurally closely related phenylpropenes (PPs), estragole, anethole, eugenol, and isoeugenol, on the fluidity of dipalmitoyl phosphatidyl choline (DPPC) liposome membrane was investigated by DSC, Raman, and fluorescence anisotropy of 1,6-diphenyl-1,3,5-hexatriene (DPH). Liposomes were prepared by thin-film hydration
Camila Fernanda Estevão-Silva et al.
Journal of natural medicines, 68(3), 567-575 (2014-05-03)
The aim of this study was to investigate the effect of anethole (AN) and eugenol (EUG) on leukocyte migration using in vitro chemotaxis and in situ microcirculation assays. BALB/c mice were used for the in vitro chemotaxis assay, and Wistar
Kyu-Sik Chang et al.
Journal of medical entomology, 51(4), 804-810 (2014-08-15)
An assessment was made of the toxicity of 12 insecticides and three essential oils as well as Bacillus thuringiensis subsp. israelensis (Bti) alone or in combination with the oil major constituents (E)-anethole (AN), (E) -cinnamaldehyde (CA), and eugenol (EU; 1:1
Matthias Onghena et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(12), 2090-2102 (2014-11-20)
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The

Articles

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service