Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34098

Supelco

Estragole

analytical standard

Synonym(s):

4-Allylanisole, p-Allylphenyl methyl ether, p-Methoxyallylbenzene, Chavicol methyl ether, Estragole, Isoanethole, Methyl chavicol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H4OCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
1099454
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.521 (lit.)

bp

215-216 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccc(CC=C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3,5-8H,1,4H2,2H3

InChI key

ZFMSMUAANRJZFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Estragole is a plant extract, used in a variety of consumer products including flavorings, perfumes and homeopathic remedies. This compound exists as a natural constituent in a variety of plants and their essential oils, including, sweet basil, sweet fennel, tarragon and Croton zehntneri which are used in food products as flavoring agents.

Application

Estragole may be used as an analytical reference standard for the quantification of the analyte in food products, Croton zehntneri aromatic plant, pharmaceutical products, herbal teas and herbal extracts using gas chromatography coupled with flame ionization detection (GC-FID) and gas chromatography coupled to mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: pimpinella

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Estragole in Pharmaceutical Products, Herbal Teas and Herbal Extracts Using GC-FID
Ismaiel AO, et al.
Journal of Applied Pharmaceutical Science, 6(12), 144-150 (2016)
Isolation, characterization and evaluation of antimicrobial and cytotoxic activity of estragole, obtained from the essential oil of croton zehntneri (euphorbiaceae)
Andrade BCT, et al.
Anais da Academia Brasileira de Ciencias, 87(1), 173-182 (2015)
Determination of estragole, safrole and eugenol methyl ether in food products.
Siano F, et al.
Food Chemistry, 81(3), 469-475 (2003)
David Fowler et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 366(1582), 3196-3209 (2011-10-19)
This paper reports measurements of land-atmosphere fluxes of sensible and latent heat, momentum, CO(2), volatile organic compounds (VOCs), NO, NO(2), N(2)O and O(3) over a 30 m high rainforest canopy and a 12 m high oil palm plantation in the
Yuji Ishii et al.
Chemical research in toxicology, 24(4), 532-541 (2011-03-10)
Estragole (ES) is a natural constituent of several herbs and spices that acts as a carcinogen in the livers of rodents. Given that the proximal electrophilic form of ES with a reactive carbocation is generated by cytochrome P450 and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service