Skip to Content
MilliporeSigma
All Photos(2)

Documents

W284220

Sigma-Aldrich

2-Pentanone

≥99.0%, natural, FG

Synonym(s):

Methyl propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2CH3
CAS Number:
Molecular Weight:
86.13
FEMA Number:
2842
Beilstein:
506058
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
7.054
NACRES:
NA.21

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.510

vapor pressure

27 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

941 °F

expl. lim.

8.2 %

refractive index

n20/D 1.39 (lit.)

bp

101-105 °C (lit.)

mp

−78 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; woody; spicy

SMILES string

CCCC(C)=O

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3

InChI key

XNLICIUVMPYHGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service