Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

THS01131

ThalesNano CatCart® catalyst cartridge system, 70 mm L

10% Pd/C

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104500
NACRES:
NB.51

packaging

pkg of 1 ea

manufacturer/tradename

ThalesNano THS 01131

diam.

4 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • ThalesNano CatCart® system is a packaged catalyst cartridge system consisting of heterogeneous catalysts of a specific particle range.
  • CatCart® is made of a stainless-steel tube sealed on both sides.
  • A filter system at each end of the tube allows only liquid to pass through the column but prevents catalysts from coming out.
  • The reusable cartridges are compatible with the H-Cube series and Phoenix Flow Reactor systems.
  • The system offers contact-less safe handling.
CatCarts are suitable for use only with Thalesnano H-Cube reactors.

Legal Information

CatCart is a registered trademark of ThalesNano Nanotechnology Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service