Skip to Content
MilliporeSigma
All Photos(3)

Documents

205931

Sigma-Aldrich

Platinum on carbon

greener alternative

extent of labeling: 5 wt. % loading, matrix activated carbon support

Synonym(s):

Pt/C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

matrix

activated carbon support

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on carbon (5 wt.% loading, matrix activated carbon support) (Pt/C) can be used as a catalyst for a variety of applications, such as:

  • hydrosilylation reaction for the production of molecules with silicon-carbon bonds
  • direct alcohol fuel cells (DAFCs)
  • an electrocatalyst in hydrogen evolution reaction (HER)
  • formation of nanocatalyst for polymer electrolyte fuel cells
  • hydrogenation of alkenes, alkynes, nitro groups, ketones, and aromatics
  • hydrogenolysis of cyclopropanes
  • oxidation of alcohols.
Pt/C can also be used as an hydrosilylation catalyst. It can convert alkenes and alkynes to chlorosilanes, alkylsilanes, and alkoxysilanes at the temperature and pressure conditions of 100-300 oC and 30-1000 psi, respectively.It may be also used as a catalyst in methanol fuel cells.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soosan Kim et al.
Chemosphere, 248, 126043-126043 (2020-02-03)
A Pt catalyst supported on activated carbon (Pt/AC) was used for an environmentally friendly thermal treatment of food waste under an inert atmosphere (i.e., pyrolysis). This catalyst influenced the amounts of condensable hydrocarbons and noncondensable gases but not that of
Soosan Kim et al.
Journal of hazardous materials, 393, 122449-122449 (2020-03-11)
In this study, a method of disposing food waste is introduced via catalytic pyrolysis under CO2 condition. The catalyst and CO2 hindered the generation of condensable compounds, leading to enhancing non-condensable gas generation. However, they did not affect the amount
Soosan Kim et al.
Journal of hazardous materials, 392, 122464-122464 (2020-03-21)
The accumulation of industrial plastic waste in the environment is a global growing concern. Thermochemical process is a preferred method to dispose plastic waste mainly because it can reduce volume of the waste; however, the thermochemical disposal of plastic waste
Platinum supported on mesoporous carbon as cathode catalyst for direct methanol fuel cells
Bruno MM, et al.
Journal of Power Sources, 278, 458-463 (2015)
Hydrosilylation of alkynes catalyzed by platinum on carbon.
Chauhan M, et al.
Journal of Organometallic Chemistry, 645(1), 1-13 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service