Key Documents
TEM-G200NI
Sigma-Aldrich
Glider Grids Standard Nickel Square Mesh
size 200 mesh, vial of 100
Synonym(s):
SQUARE 200 NESH NICKEL 100 VIAL
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
vial of 100
size
200 mesh
General description
Gilder Grids Standard Square Mesh: Nickel Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm
Choose from one of the most recent versions:
Certificates of Analysis (COA)