Key Documents
TEM-G1000HHCU
Sigma-Aldrich
1000 Mesh Hexagonal Copper Grid
vial of 25
Synonym(s):
G1000HH-CU HEXAGONAL MESH
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
vial of 25
General description
High Mesh Value Standard Hexagonal Mesh: Copper Grid, 1000 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 25 μm, hole ~19μm, bar ~6μm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)