Documents
SYNV163024
Synthware vacuum trap with serrated hose connections
joint: ST/NS 24/40, body O.D. 30 mm, L 200 mm , below joint
Synonym(s):
Synthware V163024
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V163024
L
200 mm , below joint
body O.D.
30 mm
General description
With joints for easy removal of condensate. Serrated hose connections for side tube have an O.D. of 13 mm at the smallest serration and 16 mm at the largest serration. Center tube hose connection is always 10mm for easy connection to manifold bank tubes which are also with 10 mm hose connection.
Legal Information
Synthware is a trademark of Kemtech America Inc.