Documents
Z324019
BRAND® reservoir bottle for automatic burettes
capacity 2 L, clear soda-lime glass
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
clear soda-lime glass
round bottle
Joint
joint 29/32 standard ground joint
joint ST/NS neck29/32
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 126965
capacity
2 L
diam. × H
160 mm × 200 mm
General description
DURAN® spare reservoir bottles for automatic burettes.
Other Notes
BR126965 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
DURAN is a registered trademark of DWK Life Sciences