Key Documents
SYNF449900
Synthware three neck round bottom flask with vertical side necks
100 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Synonym(s):
Synthware F449900, 3-neck flask, flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
three-neck (vertical side necks)
Joint
center joint: ST/NS19/22
side joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F449900
capacity
100 mL
100 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.