Key Documents
SYNF444400
Synthware three neck round bottom flask with vertical side necks
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(s):
Synthware F444400, 3-neck round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
three-neck (vertical side necks)
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F444400
capacity
100 mL
100 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.