Key Documents
SYNA532440
Synthware inlet or thermometer adapter with hose connection
joint: ST/NS 24/40, O.D. 5.0-7.5 mm
Synonym(s):
Synthware A532440, thermometer inlet adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A532440
O.D.
5.0-7.5 mm
Related Categories
General description
Same as thermometer adapter but with 8mm or 10mm hose connection for N2/Ar introduction or vacuum. Supplied with one Nylon cap and one Viton® O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)