Documents
SYNC264275
Synthware coil-style reflux condenser
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C264275, coil-type condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C264275
hose size
10 mm
General description
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 8mm or 10mm at the middle serration.
Legal Information
Synthware is a trademark of Kemtech America Inc.