CLS4320C1L
Pyrex® round-bottom heavy wall boiling flask, short neck with joint
capacity 1 L, joint: ST/NS 24/40
Synonym(s):
Corning® 4320C
All Photos(1)
About This Item
UNSPSC Code:
41121804
Recommended Products
material
round bottom flask
Joint
joint: ST/NS24/40
packaging
case of 1 ea
manufacturer/tradename
Corning 4320C-1L
capacity
1 L
neck size
(1)
Related Categories
General description
Pyrex heavy duty boiling flask provides increased mechanical strength. The flask features walls that are 25-30% thicker than the standard version.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)