Key Documents
SYNC164250
Synthware vacuum jacketed distillation column with indents to support packing material
joint: ST/NS 24/40, jacket H 250 mm
Synonym(s):
Synthware 164250, glass distilling column
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C164250
jacket H
250 mm
General description
With 12 mm I.D. indentations to support packing material. Vacuum jacketed to improve performance .
Legal Information
Synthware is a trademark of Kemtech America Inc.