Key Documents
SYNC109130
Synthware Vigreux distillation column
joint: ST/NS 19/22, L 130 mm
Synonym(s):
Synthware C109130, distilling column, fractional column
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C109130
L
130 mm
General description
Column length is measured from the highest to lowest indent in the column.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)